Iodobenzene, 98%
Iodobenzene, 98%
Iodobenzene, 98%
Thermo Scientific Chemicals

Iodobenzene, 98%

CAS: 591-50-4 | C6H5I | 204.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 122335000
Price (USD)
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS591-50-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.6180 to 1.6200 (20°C, 589 nm)
Appearance (Color)Clear yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only