2-Mercaptothiazoline, 98%
2-Mercaptothiazoline, 98%
2-Mercaptothiazoline, 98%
Thermo Scientific Chemicals

2-Mercaptothiazoline, 98%

CAS: 96-53-7 | C3H5NS2 | 119.21 g/mol
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 125561000
Price (JPY)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS96-53-7
IUPAC Name1,3-thiazolidine-2-thione
Molecular FormulaC3H5NS2
InChI KeyWGJCBBASTRWVJL-UHFFFAOYSA-N
SMILESS=C1NCCS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration with TBAH97.5 to 102.0 %
Melting point103°C to 106°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only