Mesitylene, 99%, Extra Pure
Mesitylene, 99%, Extra Pure
Mesitylene, 99%, Extra Pure
Thermo Scientific Chemicals

Mesitylene, 99%, Extra Pure

CAS: 108-67-8 | C9H12 | 120.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 125580010
Price (USD)
178.65
Special offer
Online exclusive
Ends: 28-Dec-2026
210.00
Save 31.35
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
178.65
Special offer
Online exclusive
Ends: 28-Dec-2026
210.00
Save 31.35
Each
Chemical Identifiers
CAS108-67-8
IUPAC Name1,3,5-trimethylbenzene
Molecular FormulaC9H12
InChI KeyAUHZEENZYGFFBQ-UHFFFAOYSA-N
SMILESCC1=CC(C)=CC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4980 to 1.5000 (20°C, 589 nm)
Appearance (Color)Clear colorless
GC>=98.5 %
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only