4-Methoxybenzophenone, 97%
4-Methoxybenzophenone, 97%
4-Methoxybenzophenone, 97%
Thermo Scientific Chemicals

4-Methoxybenzophenone, 97%

CAS: 611-94-9 | C14H12O2 | 212.25 g/mol
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 125740250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS611-94-9
IUPAC Name(4-methoxyphenyl)(phenyl)methanone
Molecular FormulaC14H12O2
InChI KeySWFHGTMLYIBPPA-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Melting point58°C to 63°C
Appearance (Form)Crystalline solid
Infrared spectrumConforms
Appearance (Color)White to yellow to light orange
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only