2'-Methylacetophenone, 98%
2'-Methylacetophenone, 98%
2'-Methylacetophenone, 98%
Thermo Scientific Chemicals

2'-Methylacetophenone, 98%

CAS: 577-16-2 | C9H10O | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 126160250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS577-16-2
IUPAC Name1-(2-methylphenyl)ethan-1-one
Molecular FormulaC9H10O
InChI KeyYXWWHNCQZBVZPV-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.5290 to 1.5330 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only