2-Methylanisole, 99%
2-Methylanisole, 99%
2-Methylanisole, 99%
Thermo Scientific Chemicals

2-Methylanisole, 99%

CAS: 578-58-5 | C8H10O | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 126281000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS578-58-5
IUPAC Name1-methoxy-2-methylbenzene
Molecular FormulaC8H10O
InChI KeyDTFKRVXLBCAIOZ-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.5160 to 1.5180 (20°C, 589 nm)
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only