2-Methylpiperazine, 98%
2-Methylpiperazine, 98%
2-Methylpiperazine, 98%
Thermo Scientific Chemicals

2-Methylpiperazine, 98%

CAS: 109-07-9 | C5H12N2 | 100.16 g/mol
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 127475000
Price (JPY)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS109-07-9
IUPAC Name2-methylpiperazine
Molecular FormulaC5H12N2
InChI KeyJOMNTHCQHJPVAZ-UHFFFAOYNA-N
SMILESCC1CNCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<1 %
Appearance (Form)Crystalline powder or crystals and/or chunks
Appearance (Color)White to yellow
Infrared spectrumConforms
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only