3-Methylpyrazole, 99%
3-Methylpyrazole, 99%
3-Methylpyrazole, 99%
Thermo Scientific Chemicals

3-Methylpyrazole, 99%

CAS: 1453-58-3 | C4H6N2 | 82.11 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 127560050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1453-58-3
IUPAC Name5-methyl-1H-pyrazole
Molecular FormulaC4H6N2
InChI KeyXKVUYEYANWFIJX-UHFFFAOYSA-N
SMILESCC1=CC=NN1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4945 to 1.4965 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to light orange
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only