3-Nitrobenzaldehyde, 99%
3-Nitrobenzaldehyde, 99%
3-Nitrobenzaldehyde, 99%
Thermo Scientific Chemicals

3-Nitrobenzaldehyde, 99%

CAS: 99-61-6 | C7H5NO3 | 151.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 128411000
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.50
Save 5.85
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.50
Save 5.85
Each
Chemical Identifiers
CAS99-61-6
IUPAC Name3-nitrobenzaldehyde
Molecular FormulaC7H5NO3
InChI KeyZETIVVHRRQLWFW-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-brown
Appearance (Form)Granular powder
Infrared spectrumConforms
Melting point56°C to 59°C
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only