Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MDSGSSSSD SAPDCWDQVD MESTGSAPSG DGVSSAVAEA QREPLSSAFS RKLNVNAKPF VPNVHAAEFV PSFLRGPTQP PTLPAGSGSN DETCTGAGYP QGKRMGRGAP VEPSREEPLV SLEGSNSAVT MELSEPVVEN GEVEMALEES WEHSKEVSEA EPGGGSSGDS GPPEESGQEM MEEKEEIRKS KSVIVPSGAP K (1-200 aa encoded by BC036077)
ERF3B encodes a GTPase that belongs to the GTP-binding elongation factor family. The encoded protein is a polypeptide release factor that complexes with eukaryotic peptide chain release factor 1 to mediate translation termination. This protein may also be involved in mRNA stability.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eRF3b; Eukaryotic peptide chain release factor GTP-binding subunit ERF3B; Eukaryotic peptide chain release factor subunit 3b; FLJ10441; G1 to S phase transition protein 2 homolog; unnamed protein product
Gene Aliases: ERF3B; GSPT2; GST2; RGD1563213
UniProt ID: (Human) Q8IYD1, (Mouse) Q149F3
Entrez Gene ID: (Human) 23708, (Mouse) 14853, (Rat) 501582
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support