Pentafluorobenzoic acid, 99%
Pentafluorobenzoic acid, 99%
Pentafluorobenzoic acid, 99%
Thermo Scientific Chemicals

Pentafluorobenzoic acid, 99%

CAS: 602-94-8 | C7HF5O2 | 212.08 g/mol
Quantity:
5 g
25 g
100 g
Catalog number 129911000
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS602-94-8
IUPAC Namepentafluorobenzoic acid
Molecular FormulaC7HF5O2
InChI KeyYZERDTREOUSUHF-UHFFFAOYSA-N
SMILESOC(=O)C1=C(F)C(F)=C(F)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystalline powder
Melting point100°C to 104°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only