1,2-Phthalic dicarboxaldehyde, 98+%
1,2-Phthalic dicarboxaldehyde, 98+%
1,2-Phthalic dicarboxaldehyde, 98+%
Thermo Scientific Chemicals

1,2-Phthalic dicarboxaldehyde, 98+%

CAS: 643-79-8 | C8H6O2 | 134.13 g/mol
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 131080250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS643-79-8
IUPAC Namebenzene-1,2-dicarbaldehyde
Molecular FormulaC8H6O2
InChI KeyZWLUXSQADUDCSB-UHFFFAOYSA-N
SMILESO=CC1=CC=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or lumps
Appearance (Color)Yellow
Infrared spectrumConforms
Melting point54°C to 58°C
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only