Product Image
Thermo Scientific Chemicals

Propionic anhydride, 99

CAS: 123-62-6 | C6;H1;0;O3; | 130.14 g/mol
Quantity:
250 mL
1 L
2.5 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 131520025
Price (JPY)
-
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS123-62-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<20 APHA
Infrared spectrumConforms
Refractive index1.4030 to 1.4050 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only