D(-)-Ribose, 99+%
D(-)-Ribose, 99+%
D(-)-Ribose, 99+%
Thermo Scientific Chemicals

D(-)-Ribose, 99+%

CAS: 50-69-1 | C5H10O5 | 150.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 132360250
Price (USD)
54.30
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
54.30
Each
Chemical Identifiers
CAS50-69-1
IUPAC Name(3R,4S,5R)-5-(hydroxymethyl)oxolane-2,3,4-triol
Molecular FormulaC5H10O5
InChI KeyHMFHBZSHGGEWLO-ZHZWXSSZNA-N
SMILESOC[C@H]1OC(O)[C@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<1 % (60°C, 3 h)
Specific optical rotation-18° to -22° (20°C, 589 nm) (c=2, water)
Appearance (Color)White to light beige
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only