DL-Serine, 99%
DL-Serine, 99%
DL-Serine, 99%
Thermo Scientific Chemicals

DL-Serine, 99%

CAS: 302-84-1 | C3H7NO3 | 105.093 g/mol
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 132651000
Price (JPY)
-
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS302-84-1
IUPAC Name2-amino-3-hydroxypropanoic acid
Molecular FormulaC3H7NO3
InChI KeyMTCFGRXMJLQNBG-UHFFFAOYNA-N
SMILESNC(CO)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Other amino acids=<0.5 % (TLC)
Heavy metals (as Pb)=<10 ppm
Iron (Fe)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only