beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol
Thermo Scientific Chemicals

beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol

CAS: 83-46-5 | C29H50O | 414.718 g/mol
Catalog NumberQuantityPackaging
132725000500 gPlastic bottle
Catalog number 132725000
Price (JPY)
-
Request A Quote
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS83-46-5
IUPAC Name(1R,3aS,3bS,7S,9aR,9bS,11aR)-1-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Molecular FormulaC29H50O
InChI KeyKZJWDPNRJALLNS-VJSFXXLFSA-N
SMILESCC[C@H](CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<3.5 %
GC>=95 % Total sterols
GC10 to 14 % (Beta-sitostanol) Typical
Appearance (Form)Powder
Specific optical rotation-24° to -32° (20°C, 589 nm) (c=2, CHCl3)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only