2-Butyne, 98%, Thermo Scientific Chemicals
2-Butyne, 98%, Thermo Scientific Chemicals
2-Butyne, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Butyne, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
1351400505 gGlass bottle
Catalog number 135140050
Price (USD)
62.70
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
62.70
Each
Chemical Identifiers
CAS503-17-3
IUPAC Namebut-2-yne
Molecular FormulaC4H6
InChI KeyXNMQEEKYCVKGBD-UHFFFAOYSA-N
SMILESCC#CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.3920 to 1.3940 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only