2,2,2-Trichloroethanol, 99%
2,2,2-Trichloroethanol, 99%
2,2,2-Trichloroethanol, 99%
Thermo Scientific Chemicals

2,2,2-Trichloroethanol, 99%

CAS: 115-20-8 | C2H3Cl3O | 149.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 139441000
Price (EUR)
43,65
Online exclusive
47,90
Save 4,25 (9%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
43,65
Online exclusive
47,90
Save 4,25 (9%)
Each
Chemical Identifiers
CAS115-20-8
IUPAC Name2,2,2-trichloroethan-1-ol
Molecular FormulaC2H3Cl3O
InChI KeyKPWDGTGXUYRARH-UHFFFAOYSA-N
SMILESOCC(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4880 to 1.4910 (20°C, 589 nm)
GC>=98.5 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only