Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • IL-17A Antibodies
          Now also available in recombinant format (Product # 740005M) and additional conjugated formats in both full and sample sizes. Search using the clone ID "eBio64DEC17" or explore the "Additional formats" section on this page.

          Invitrogen

          IL-17A Monoclonal Antibody (eBio64DEC17), eBioscience™

          55 Published Figures
          71 References
          View all (193) IL-17A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite IL-17A Monoclonal Antibody (eBio64DEC17), eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Published Figures (55)
          IL-17A Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          IL-17A Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 55

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IL-17A Antibody (14-7179-37) in IHC

          The Journal of clinical investigation 2023 - In vivo administration of rhCTRP4 attenuates clinical severity. (A) After EAE induction of WT mice, 500 ng/mice rhCTRP4 or control (BSA) were administered intraperitoneally every day starting at disease onset on day 9 after immunization until sacrificed. The clinical scores were monitored daily and are depicted (n = 10/group). (B) Representative images of H&E and LFB staining on spinal cord sections of mice... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IL-17A Antibody in Immunohistochemistry (IHC)
          IL-17A Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          IL-17A Antibody in Immunocytochemistry, Radioimmune assays (ICC/IF, RIA)
          IL-17A Antibody in Immunocytochemistry (ICC/IF)
          IL-17A Antibody in Immunocytochemistry (ICC/IF)
          IL-17A Antibody in Immunohistochemistry (IHC)
          IL-17A Antibody in Immunohistochemistry (IHC)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in Flow Cytometry (Flow)
          IL-17A Antibody in ELISA (ELISA)
          IL-17A Antibody in ELISA (ELISA)
          IL-17A Antibody in ELISA (ELISA)
          IL-17A Antibody in ELISA (ELISA)
          IL-17A Antibody in ELISA (ELISA)
          IL-17A Antibody in ELISA (ELISA)
          IL-17A Antibody in Neutralization (Neu)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          14-7179-37

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-Dependent
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 7 publications 7 publications

          Flow Cytometry (Flow)

          -
          View 46 publications 46 publications

          ELISA (ELISA)

          Assay-Dependent
          View 5 publications 5 publications

          Neutralization (Neu)

          -
          View 3 publications 3 publications

          Radioimmune Assays (RIA)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human

          Published species

          Hamster, Human, Mouse, Non-human primate, Rat, Rhesus monkey, Sheep

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBio64DEC17

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 563
          • Brilliant UV 737
          • Brilliant UV 805
          • Brilliant Violet 605
          • Brilliant Violet 786
          • eFluor 450
          • eFluor 506
          • eFluor 660
          • FITC
          • PE 
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-Cyanine5.5
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          2 mg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2865271

          Product Specific Information

          Description: The eBio64DEC17 antibody reacts with human IL-17A. The eBio64DEC17 antibody is a neutralizing antibody. Interleukin-17A (IL-17A) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. The cDNA encoding human IL-17A was isolated from a library of CD4+ T cells; the encoded protein exhibits 72 percent amino acid identity with HVS13 , an open reading frame from a T lymphotropic Herpesvirus saimiri, and 63 percent with mouse CTLA-8 (cytotoxic T-lymphocyte associated antigen-8). Human IL-17A exists as glycosylated 20-30 kD homodimers. High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells. IL-17A enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. Human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse IL-17A receptor.



          IL-23-dependent, IL-17A-producing CD4+ T cells (Th-17 cells) have been identified as a unique subset of Th cells that develops along a pathway that is distinct from the Th1- and Th2- cell differentiation pathways. The hallmark effector molecules of Th1 and Th2 cells, e.g., IFN gamma and IL-4, have each been found to negatively regulate the generation of these Th-17 cells.

          Intracellular staining by eBio64DEC17 antibody identifies the same cell population as the eBio64CAP17 antibody, as can be seen in co-staining experiments using both antibodies.

          Applications Reported: This eBio64DEC17 antibody has been reported for use in immunohistology staining and ELISA.

          Applications Tested: This eBio64DEC17 antibody has been tested by ELISA.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Interleukin-17A (IL-17A, CTLA-8) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. IL-17A is a 32 kDa long, disulfide-linked homodimer consisting of 136 amino acids that is a member of a six-species family of proteins (IL-17A-17F) and signals through the IL-17 receptor (IL-17R/CDw217). High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells, and IL-17A also enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. In particular, human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A regulates the activities of NF-kappa B and mitogen-activated protein kinases, stimulates the expression of IL-6 and cyclooxygenase-2 (PTGS2/COX-2), and enhances the production of nitric oxide (NO).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; ILN; inflammatory cytokine; Interleukin; interleukin 17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin-17; Interleukin17A; R-IL-17-A

          View more View less

          Gene Aliases: CTLA-8; CTLA8; IL-17; IL-17A; IL17; ILA17

          View more View less

          UniProt ID: (Human) Q16552

          View more View less

          Entrez Gene ID: (Human) 3605

          View more View less

          Function(s)
          cytokine activity protein binding protein homodimerization activity protein heterodimerization activity
          Process(es)
          positive regulation of antimicrobial peptide production apoptotic process inflammatory response immune response Notch signaling pathway cell-cell signaling cell death response to wounding gene expression keratinocyte differentiation positive regulation of interleukin-1 beta production positive regulation of interleukin-12 production positive regulation of interleukin-16 production positive regulation of interleukin-23 production positive regulation of interleukin-6 production positive regulation of tumor necrosis factor production interleukin-17A-mediated signaling pathway keratinocyte proliferation positive regulation of osteoclast differentiation positive regulation of transcription from RNA polymerase II promoter defense response to Gram-negative bacterium defense response to Gram-positive bacterium defense response to fungus intestinal epithelial structure maintenance protein K63-linked ubiquitination fibroblast activation T-helper 17 type immune response interleukin-17-mediated signaling pathway granulocyte migration negative regulation of inflammatory response to wounding positive regulation of cytokine production involved in inflammatory response positive regulation of bicellular tight junction assembly positive regulation of tumor necrosis factor superfamily cytokine production positive regulation of chemokine (C-X-C motif) ligand 1 production
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-7d6nq:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline