Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PD-1 (CD279) Antibodies

          Invitrogen

          CD279 (PD-1) Monoclonal Antibody (J43), eBioscience™

          42 Published Figures
          80 References
          View all (391) PD-1 (CD279) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD279 (PD-1) Monoclonal Antibody (J43), eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (42)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 43

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD279 (PD-1) Antibody (14-9985-82) in Flow

          Staining of 3-day unstimulated (left) and 3-day ConA activated (right) BALB/c splenocytes with 0.25 µg of Hamster IgG Isotype Control Purified (Product # 14-4888-81) (open histogram) or 0.25 µg of Anti-Mouse PD-1 Purified (filled histogram) followed by Anti-Armenian Hamster IgG Biotin (Product # 13-4113-85) and Streptavidin PE (Product # 12-4317-87).Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)
          CD279 (PD-1) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          14-9985-82

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-Dependent
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          0.5 µg/test
          View 71 publications 71 publications

          Immunoprecipitation (IP)

          Assay-Dependent
          -

          Neutralization (Neu)

          -
          View 1 publication 1 publication

          Functional Assay (Functional)

          -
          View 3 publications 3 publications

          Inhibition Assays (Inhibition)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Mouse

          Host/Isotype

          Armenian hamster / IgG

          Class

          Monoclonal

          Type

          Antibody

          Clone

          J43

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 563
          • Brilliant UV 615
          • Brilliant UV 661
          • Brilliant UV 737
          • Brilliant UV 805
          • Brilliant Violet 480
          • Brilliant Violet 605
          • Brilliant Violet 650
          • Brilliant Violet 786
          • eFluor 450
          • eFluor 506
          • FITC
          • Functional Grade
          • PE 
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Super Bright 702
          • Super Bright 780
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_468664

          Product Specific Information

          Description: The J43 monoclonal antibody reacts with mouse PD-1 (programmed death-1), a 55 kDa member of the Ig superfamily. PD-1 contains the immunoreceptor tyrosine-based inhibitory motif (ITIM) and plays a key role in peripheral tolerance and autoimmune disease in mice. PD-1 is expressed mainly on activated T and B lymphocytes. Two novel B7 Family members have been identified as PD-1 ligands, PD-L1 (B7-H1) and PD-L2 (B7-DC). Evidence reported to date suggests overlapping functions for these ligands and their constitutive expression on some normal tissues and upregulation on activated antigen-presenting cells. It is reported that J43 inhibits the binding of mouse PD-L1-Ig and mouse PD-L2-Ig to PD-1/BHK transfected cells. When administrated in vivo, both intact and Fab of J43 are reported to enhance contact hypersensitivity and exacerbate acute GVHD similar to transfer of PD-1-deficient cells. Injection of J43 also exacerbates EAE and NOD diabetes as do specific antibodies to mouse PD-L1 and PD-L2.

          Applications Reported: The J43 antibody has been reported for use in flow cytometric analysis, and immunoprecipitation. It has also been reported for use in in vitro functional assays. (Please use Functional Grade purified J43, Product # 16-9985, in functional assays).

          Applications Tested: The J43 antibody has been tested by flow cytometric analysis of mouse Con-A activated spleen cells and mouse PD-1 transfected cells. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Cell-mediated immune responses are initiated by T lymphocytes that are themselves stimulated by cognate peptides bound to MHC molecules on antig en-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as PD-1 (also known as PDCD-1) that mediate inhibitory signals from the APC. PD-1 can bind two different but related ligands, PDL-1 and PDL-2. Upon binding to either of these ligands, signals generated by PD-1 inhibit the activation of the immune response in the absence of "danger signals" such as LPS or other molecules associated with bacteria or other pathogens. Evidence for this is seen in PD1-null mice who exhibit hyperactivated immune systems and autoimmune diseases. Despite its predicted molecular weight, PD-1 often migrates at higher molecular weight in SDS-PAGE.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CD279; PD-1; hPD1; mPD-1; PD-1; programmed cell death 1 protein; protein PD-1; Protein PD1; sCD279; soluble CD279; unnamed protein product

          View more View less

          Gene Aliases: Ly101; PD-1; Pdc1

          View more View less

          UniProt ID: (Mouse) Q02242

          View more View less

          Entrez Gene ID: (Mouse) 18566

          View more View less

          Function(s)
          transmembrane signaling receptor activity protein binding signaling receptor activity
          Process(es)
          negative regulation of tolerance induction negative regulation of T cell mediated immune response to tumor cell positive regulation of apoptotic process negative regulation of apoptotic process regulation of immune response negative regulation of immune response negative regulation of T cell receptor signaling pathway negative regulation of T cell activation negative regulation of multicellular organismal process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nv6df:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline