2-Hexanone, 98%
2-Hexanone, 98%
2-Hexanone, 98%
Thermo Scientific Chemicals

2-Hexanone, 98%

2-Hexanone, 98%, C6H12O, CAS Number-591-78-6, 2-hexanone, 2-oxohexane, butyl methyl ketone, hexanone, hexanone-2, ketone, butyl methyl, methyl butyl ketone, methyl n-butyl ketone, n-butyl methyl ketone, propylacetone, 500mL, 01, 689, 127 deg.C, 100.0, 97.5, Colorless to Yellow, 0.8100g/mL, 209-731-1 | CAS: 591-78-6 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 146881000
Price (USD)
119.65
Special offer
Online exclusive
Ends: 28-Dec-2026
140.00
Save 20.35
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
119.65
Special offer
Online exclusive
Ends: 28-Dec-2026
140.00
Save 20.35
Each
Chemical Identifiers
CAS591-78-6
IUPAC Namehexan-2-one
Molecular FormulaC6H12O
InChI KeyQQZOPKMRPOGIEB-UHFFFAOYSA-N
SMILESCCCCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.3995 to 1.4015 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only