Trifluoroacetic anhydride, 99+%
Trifluoroacetic anhydride, 99+%
Trifluoroacetic anhydride, 99+%
Thermo Scientific Chemicals

Trifluoroacetic anhydride, 99+%

Trifluoroacetic anhydride, 99+%, Quantity: 500g, Packaging: Glass Bottle, Boiling Point: 39.1 deg.C, Undesignated, Melting Point: -63.5 deg.C, Molecular Weight: 210.03, Percent Purity: 99+%, Assay Percent Range: 99% min. (GC), Beilstein: 02, II, 186, CAS: 407-25-0 | CAS: 407-25-0 | C4F6O3 | 210.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 147812501
Price (USD)
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS407-25-0
IUPAC Name2,2,2-trifluoroacetyl 2,2,2-trifluoroacetate
Molecular FormulaC4F6O3
InChI KeyQAEDZJGFFMLHHQ-UHFFFAOYSA-N
SMILESFC(F)(F)C(=O)OC(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %
Sulfate (SO4)=<500 ppm
Fluoride (F)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent
RUO – Research Use Only