1,2-Bis(diphenylphosphino)ethane, 98+%
1,2-Bis(diphenylphosphino)ethane, 98+%
1,2-Bis(diphenylphosphino)ethane, 98+%
Thermo Scientific Chemicals

1,2-Bis(diphenylphosphino)ethane, 98+%

CAS: 1663-45-2 | C26H24P2 | 398.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 147910500
Price (USD)
324.65
Online exclusive
361.00
Save 36.35
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
324.65
Online exclusive
361.00
Save 36.35
Each
Chemical Identifiers
CAS1663-45-2
IUPAC Name[2-(diphenylphosphanyl)ethyl]diphenylphosphane
Molecular FormulaC26H24P2
InChI KeyQFMZQPDHXULLKC-UHFFFAOYSA-N
SMILESC(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Melting point138°C to 144°C
HPLC>=98.0 %
Appearance (Color)White to off-white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only