Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Ly-6A/E (Sca-1) Antibodies

          Invitrogen

          Ly-6A/E (Sca-1) Monoclonal Antibody (D7), PE-Cyanine5, eBioscience™

          98 Published Figures
          115 References
          View all (35) Ly-6A/E (Sca-1) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Ly-6A/E (Sca-1) Monoclonal Antibody (D7), PE-Cyanine5, eBioscience™

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (98)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 99

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Ly-6A/E (Sca-1) Antibody (15-5981-82) in Flow

          Staining of C57Bl/6 splenocytes with Anti-Mouse CD3e FITC (Product # 11-0031-82) and 0.03 µg of Anti-Mouse Ly-6A/E (Sca-1) PE-Cyanine5.Total viable cells were used for analysis.Quadrants were set based on the autofluorescence sample. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)
          Ly-6A/E (Sca-1) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          15-5981-82

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.125 µg/test
          View 115 publications 115 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Fish, Human, Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), PE-Cyanine5, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          D7

          Conjugate

          PE-Cyanine5 PE-Cyanine5 PE-Cyanine5
          • Unconjugated
          • Alexa Fluor 700 
          • APC 
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 661
          • Brilliant UV 737
          • Brilliant UV 805
          • Brilliant Violet 421
          • Brilliant Violet 480
          • Brilliant Violet 605
          • Brilliant Violet 711
          • Brilliant Violet 786
          • FITC
          • PE 
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-Cyanine5.5
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Request custom conjugation

          Excitation/Emission Max

          568/666 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_468819

          Product Specific Information

          Description: The D7 monoclonal antibody reacts with mouse Sca-1, an 18 kDa member of the Ly-6 family of GPI-linked surface proteins. D7 reacts with both Ly-6E.1 and Ly-6A.2 molecules expressed by mouse hematopoietic stem cells, myeloid population and peripheral T and B cells. Non-activated splenocytes from Ly-6.2 strains including C57BL, SJL, 129, AKR, and others demonstrate higher frequency of D7-positive cells compared to the Ly-6.1 strains such as BALB/c, C3H, NZB and others. Sca-1 is expressed at high levels upon activation regardless of the Ly-6 haplotype. The Ly-6 family is involved in regulation and function of T cell activation. Sca-1 is a major phenotypic marker for mouse hematopoietic progenitor/stem cell subset.

          Applications Reported: This D7 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This D7 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light.

          Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-822-49) (100 µL cell sample + 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-54) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

          Excitation: 488-561 nm; Emission: 667 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Stem Cell Antigen-1 (Sca-1) is a member of the lymphocyte antigen 6 (Ly-6) family. It is expressed on multipotent hematopoietic stem cells (HSC) and is used as a marker of HSC in mice. Sca-1 is used in combination with lineage depletion antibodies to identify murine HSC. Sca-1 can be found in fetal liver, adult bone marrow, and adult peripheral blood and spleen. Sca-1 may be involved in the regulation of B and T cell activation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: locus A/E; Locus AE; ly6a-e; Ly6A/E; Ly6e; lymphocyte antigen 6 complex; lymphocyte antigen 6 complex, locus A; lymphocyte antigen 6 complex, locus E; stem cell antigen 1; stem cell antigen 2; T-cell-activating protein; T-cell-activating protein precursor; thymic shared antigen 1; unnamed protein product

          View more View less

          Gene Aliases: Ly-6A.2; Ly-6A/E; Ly-6E.1; Ly67; RIG-E; Sca-1; Sca-2; Sca1; TAP; TSA-1; Tsa1

          View more View less

          UniProt ID: (Mouse) P05533, (Mouse) Q64253

          View more View less

          Entrez Gene ID: (Mouse) 110454, (Mouse) 17069

          View more View less

          Function(s)
          protein binding acetylcholine receptor inhibitor activity acetylcholine receptor binding
          Process(es)
          in utero embryonic development adrenal gland development organ growth norepinephrine metabolic process negative regulation of viral entry into host cell epinephrine secretion ventricular cardiac muscle tissue morphogenesis acetylcholine receptor signaling pathway response to bacterium
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-6qggw:80/100.66.76.149:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline