Diethyl succinate, 98%
Diethyl succinate, 98%
Diethyl succinate, 98%
Thermo Scientific Chemicals

Diethyl succinate, 98%

CAS: 123-25-1 | C8H14O4 | 174.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
25 kg
Packaging:
View all
Plastic drum
Catalog number 150540051
Price (USD)
256.00
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
256.00
Each
Chemical Identifiers
CAS123-25-1
IUPAC Name1,4-diethyl butanedioate
Molecular FormulaC8H14O4
InChI KeyDKMROQRQHGEIOW-UHFFFAOYSA-N
SMILESCCOC(=O)CCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.4180 to 1.4220 (20°C, 589 nm)
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only