Acetyl chloride, 98%
Acetyl chloride, 98%
Acetyl chloride, 98%
Thermo Scientific Chemicals

Acetyl chloride, 98%

CAS: 75-36-5 | C2H3ClO | 78.50 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 151270025
Price (ZAR)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS75-36-5
IUPAC Nameacetyl chloride
Molecular FormulaC2H3ClO
InChI KeyWETWJCDKMRHUPV-UHFFFAOYSA-N
SMILESCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.3876 to 1.3896 (20°C, 589 nm)
Color scale=<30 APHA
Appearance (Form)Clear liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only