1500 Series Class II Type B2 Bio Safety Cabinets
Thermo Scientific™

1500 Series Class II Type B2 Bio Safety Cabinets

Our Class II, Type B2 Biological Safety Cabinet is designed for applications that require greater dilution of volatile toxic chemicals or the aseptic processing of hazardous drugs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPlug TypeWidth (English) CabinetDimensions (L x W x H) ExteriorLighting
1512B2North America (115 V, 15 A)6 ft.31.6 x 78 x 72.6 in.Without UV light
1501B2North America (115 V, 15 A)4 ft.31.6 x 54.3 x 72.6 in.Without UV light
1513B2North America (230 V, NEMA 6-15)4 ft.31.6 x 54.3 x 72.6 in.Without UV light
1524B2North America (230 V, NEMA 6-15)6 ft.31.6 x 78 x 72.6 in.Without UV light
1513B2FSchuko (230 V)4 ft.31.6 x 54.3 x 72.6 in.Without UV light
1524B2FSchuko (230 V)6 ft.31.6 x 78 x 72.6 in.Without UV light
1513B2GUK (230 V)4 ft.31.6 x 54.3 x 72.6 in.Without UV light
1524B2GUK (230 V)6 ft.31.6 x 78 x 72.6 in.Without UV light
1502B2North America (115 V, 15 A)4 ft.31.6 x 54.3 x 72.6 in.With UV light
1519B2North America (115 V, 15 A)6 ft.31.6 x 78 x 72.6 in.With UV light
1514B2North America (230 V, NEMA 6-15)4 ft.31.6 x 54.3 x 72.6 in.With UV light
1529B2North America (230 V, NEMA 6-15)6 ft.31.6 x 78 x 72.6 in.With UV light
1514B2FSchuko (230 V)4 ft.31.6 x 54.3 x 72.6 in.With UV light
1529B2FSchuko (230 V)6 ft.31.6 x 78 x 72.6 in.With UV light
1514B2GUK (230 V)4 ft.31.6 x 54.3 x 72.6 in.With UV light
1529B2GUK (230 V)6 ft.31.6 x 78 x 72.6 in.With UV light
Catalog number 1512B2
Price (USD)
-
Plug Type:
North America (115 V, 15 A)
Width (English) Cabinet:
6 ft.
Dimensions (L x W x H) Exterior:
31.6 x 78 x 72.6 in.
Lighting:
Without UV light
Thermo Scientific™ 1500 Series Class II, Type B2 BSC is available in nominal widths of 4 ft (1.2 m) and 6 ft (1.8 m) and is fully compliant with NSF/ANSI 49 standards. It features an ergonomic design, advanced DC motor technology, and a convenient touchscreen user interface allows for easy monitoring of cabinet conditions.
  • NSF/ANSI 49 listed Class II, Type B2 BSC
  • Auto compensation for downflow, allowing for safe use of air pre-filters for additional extension of HEPA filter life
  • Advanced DC motor
  • Stainless steel interior
  • 5 in. touchscreen display for visualizing and monitoring cabinet conditions, including, airflow status, alerts and alarms, UV light timer, HEPA filter life indicator, Data logging, and Connectivity
Specifications
Dimensions (L x W x H) Exterior31.6 x 78 x 72.6 in.
Electrical Requirements115 V, 50/60 Hz
Energy Consumption (Operating Set Point)0.37 kW
LightingWithout UV light
Plug TypeNorth America (115 V, 15 A)
Sash Opening (English)8 in.
Width (English) Exterior78 in.
Width (English) Interior72.5 in.
Width (Metric) Exterior198.1 cm
Certifications/ComplianceNSF Class II Type B2 BSC, NSF/ANSI 49, UL, CE
Chamber MaterialStainless Steel (304)
Model1500 Series B2
Noise Level66 dBA
Product TypeBiosafety Cabinet
Receptacles2 Duplex, GFI
Sash Opening (Metric)20.3 cm
Voltage115 V
Width (English) Cabinet6 ft.
Unit SizeEach