2-Carboxybenzaldehyde, 99%
2-Carboxybenzaldehyde, 99%
2-Carboxybenzaldehyde, 99%
Thermo Scientific Chemicals

2-Carboxybenzaldehyde, 99%

CAS: 119-67-5 | C8H6O3 | 150.13 g/mol
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 152081000
Price (JPY)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS119-67-5
IUPAC Name2-formylbenzoic acid
Molecular FormulaC8H6O3
InChI KeyDYNFCHNNOHNJFG-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=98.5 %
Appearance (Form)Crystalline powder
Melting point96°C to 99°C
Appearance (Color)White to light beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only