o-Tolualdehyde, 98%, stabilized
o-Tolualdehyde, 98%, stabilized
o-Tolualdehyde, 98%, stabilized
Thermo Scientific Chemicals

o-Tolualdehyde, 98%, stabilized

CAS: 529-20-4 | C8H8O | 120.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
Catalog number 152432500
Price (USD)
347.00
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
347.00
Each
Chemical Identifiers
CAS529-20-4
IUPAC Name2-methylbenzaldehyde
Molecular FormulaC8H8O
InChI KeyBTFQKIATRPGRBS-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive index1.5460 to 1.5490 (20°C, 589 nm)
Infrared spectrumConforms
Stabilizer0.1 to 0.5 % hydroquinone
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only