m-Anisic acid, 98%
m-Anisic acid, 98%
m-Anisic acid, 98%
Thermo Scientific Chemicals

m-Anisic acid, 98%

CAS: 586-38-9 | C8H8O3 | 152.15 g/mol
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 152475000
Price (JPY)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS586-38-9
IUPAC Name3-methoxybenzoic acid
Molecular FormulaC8H8O3
InChI KeyXHQZJYCNDZAGLW-UHFFFAOYSA-N
SMILESCOC1=CC=CC(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
GC>=97.5 %
Appearance (Color)Off-white to yellow to light beige
Infrared spectrumConforms
Melting point103°C to 108°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only