2-Aminothiophenol, 98%
2-Aminothiophenol, 98%
2-Aminothiophenol, 98%
Thermo Scientific Chemicals

2-Aminothiophenol, 98%

CAS: 137-07-5 | C6H7NS | 125.19 g/mol
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 153905000
Price (JPY)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS137-07-5
IUPAC Name2-aminobenzene-1-thiol
Molecular FormulaC6H7NS
InChI KeyVRVRGVPWCUEOGV-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to orange
Infrared spectrumConforms
Appearance (Form)Liquid
Titration Argentometric97.5 to 102.5 %
Refractive index1.6410 to 1.6450 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only