Anisole, 99%, pure
Anisole, 99%, pure
Anisole, 99%, pure
Thermo Scientific Chemicals

Anisole, 99%, pure

CAS: 100-66-3 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 153920025
Price (USD)
172.65
Online Exclusive
192.00
Save 19.35 (10%)
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
172.65
Online Exclusive
192.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS100-66-3
IUPAC Nameanisole
Molecular FormulaC7H8O
InChI KeyRDOXTESZEPMUJZ-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Water=<0.1 % (K.F.)
Infrared spectrumConforms
Refractive index1.5160 to 1.5190 (20°C, 589 nm)
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only