2-Chloro-5-nitrobenzoic Acid, 99+%
2-Chloro-5-nitrobenzoic Acid, 99+%
2-Chloro-5-nitrobenzoic Acid, 99+%
Thermo Scientific Chemicals

2-Chloro-5-nitrobenzoic Acid, 99+%

CAS: 2516-96-3 | C7H4ClNO4 | 201.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 154695000
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS2516-96-3
IUPAC Name2-chloro-5-nitrobenzoic acid
Molecular FormulaC7H4ClNO4
InChI KeyQUEKGYQTRJVEQC-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=CC=C1Cl)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=99.0 %
Appearance (Form)Crystalline powder
Melting point164°C to 168°C
Infrared spectrumConforms
Appearance (Color)Light yellow to beige to light brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only