Phthalic Anhydride, 99%
Phthalic Anhydride, 99%
Phthalic Anhydride, 99%
Thermo Scientific Chemicals

Phthalic Anhydride, 99%

CAS: 85-44-9 | C8H4O3 | 148.12 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
檢視全部
Plastic bottle
產品號碼 154960025
價格 (TWD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
化學識別
CAS85-44-9
IUPAC Name1,3-dihydro-2-benzofuran-1,3-dione
Molecular FormulaC8H4O3
InChI KeyLGRFSURHDFAFJT-UHFFFAOYSA-N
SMILESO=C1OC(=O)C2=CC=CC=C12
檢視更多
規格Specification Sheet規格表
Appearance (Color)White
Appearance (Form)Flakes
Melting point130°C to 134°C
Infrared spectrumConforms
Titration by coupling98.5 to 101.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only