3-Aminobenzoic acid, 99+%
3-Aminobenzoic acid, 99+%
3-Aminobenzoic acid, 99+%
Thermo Scientific Chemicals

3-Aminobenzoic acid, 99+%

CAS: 99-05-8 | C7H7NO2 | 137.14 g/mol
Quantity:
25 g
100 g
500 g
Catalog number 155851000
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS99-05-8
IUPAC Name3-aminobenzoic acid
Molecular FormulaC7H7NO2
InChI KeyXFDUHJPVQKIXHO-UHFFFAOYSA-N
SMILESNC1=CC=CC(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=99.0 %
Appearance (Form)Powder
Melting point>=172°C
Infrared spectrumConforms
Appearance (Color)White to beige to light brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only