trans-2-Hexen-1-ol, 96%
trans-2-Hexen-1-ol, 96%
trans-2-Hexen-1-ol, 96%
Thermo Scientific Chemicals

trans-2-Hexen-1-ol, 96%

CAS: 928-95-0 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 158140250
Price (USD)
32.65
Online exclusive
36.20
Save 3.55
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
32.65
Online exclusive
36.20
Save 3.55
Each
Chemical Identifiers
CAS928-95-0
IUPAC Name(2E)-hex-2-en-1-ol
Molecular FormulaC6H12O
InChI KeyZCHHRLHTBGRGOT-SNAWJCMRSA-N
SMILESCCC\C=C\CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=95 %
Refractive index1.4350 to 1.4410 (20°C, 589 nm)
Specific gravity(20°C) 0.841 to 0.848
Appearance (Color)Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only