Thiazolyl blue tetrazolium bromide, 98%
Thiazolyl blue tetrazolium bromide, 98%
Thiazolyl blue tetrazolium bromide, 98%
Thermo Scientific Chemicals

Thiazolyl blue tetrazolium bromide, 98%

CAS: 298-93-1 | C18H16BrN5S | 414.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number 158990010
Price (USD)
62.65
Special offer
Online exclusive
Ends: 28-Dec-2026
73.10
Save 10.45
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
62.65
Special offer
Online exclusive
Ends: 28-Dec-2026
73.10
Save 10.45
Each
Chemical Identifiers
CAS298-93-1
IUPAC Name2-(4,5-dimethyl-1,3-thiazol-2-yl)-3,5-diphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium bromide
Molecular FormulaC18H16BrN5S
InChI KeyAZKSAVLVSZKNRD-UHFFFAOYSA-M
SMILES[Br-].CC1=C(C)N=C(S1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Specific extinction E (1%/1cm)>=480 (243 nm, water)
Titration Argentometric>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only