Diisobutylamine, 99%
Diisobutylamine, 99%
Diisobutylamine, 99%
Thermo Scientific Chemicals

Diisobutylamine, 99%

CAS: 110-96-3 | C8H20N | 130.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Catalog number 159070025
Price (USD)
144.65
Online exclusive
160.00
Save 15.35
Each
Quantity:
2.5 L
Request bulk or custom format
Price (USD)
144.65
Online exclusive
160.00
Save 15.35
Each
Chemical Identifiers
CAS110-96-3
SpecificationsSpecification SheetSpecification Sheet
Color scale=<15 APHA
GC>=98.5 %
Water=<0.2 %
Refractive index1.4071 to 1.4091 (20°C, 589 nm)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only