Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CCL2 (MCP-1) Antibodies

          Invitrogen

          CCL2 (MCP-1) Monoclonal Antibody (2H5), Functional Grade, eBioscience™

          7 Published Figures
          15 References
          View all (76) CCL2 (MCP-1) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CCL2 (MCP-1) Monoclonal Antibody (2H5), Functional Grade, eBioscience™

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Published Figures (7)
          CCL2 (MCP-1) Antibody in Immunocytochemistry, Flow Cytometry, ELISA (ICC/IF, Flow, ELISA)
          Group 53 Created with Sketch.
          CCL2 (MCP-1) Antibody in Immunocytochemistry, Flow Cytometry, ELISA (ICC/IF, Flow, ELISA)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CCL2 (MCP-1) Antibody (16-7096-38) in ICC/IF, Flow, ELISA

          Frontiers in immunology 2024 - Figure 6 (A) Left, representative flow cytometry histogram of CCL2 signal in 7d ligated control and experimental mice. Middle and right, quantification of percent CCL2 + ICAM1 + fibroblasts (Lin - PDGFRA + ) and CCL2 + pericytes (Lin - CD146 + ) normalized by lineage-negative mesenchymal cell numbers. (B) Representative flow cytometry histogram of ICAM1 expression in fibroblasts pre-gated for CCL2 + signal ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CCL2 (MCP-1) Antibody in Immunocytochemistry, Flow Cytometry, ELISA (ICC/IF, Flow, ELISA)
          CCL2 (MCP-1) Antibody in Immunocytochemistry (ICC/IF)
          CCL2 (MCP-1) Antibody in Immunocytochemistry (ICC/IF)
          CCL2 (MCP-1) Antibody in Immunocytochemistry (ICC/IF)
          CCL2 (MCP-1) Antibody in Immunohistochemistry (IHC)
          CCL2 (MCP-1) Antibody in Flow Cytometry (Flow)
          CCL2 (MCP-1) Antibody in Neutralization (Neu)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          16-7096-38

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 2 publications 2 publications

          Flow Cytometry (Flow)

          -
          View 2 publications 2 publications

          ELISA (ELISA)

          -
          View 4 publications 4 publications

          Neutralization (Neu)

          Assay-Dependent
          View 3 publications 3 publications

          Functional Assay (Functional)

          Assay-Dependent
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Armenian hamster / IgG

          Recommended Isotype Control

          Armenian Hamster IgG Isotype Control (eBio299Arm), Functional Grade, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2H5

          Conjugate

          Functional Grade Functional Grade Functional Grade
          • Unconjugated
          • Biotin 
          • FITC
          • PE 
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          5 mg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2866227

          Product Specific Information

          Description: The 2H5 monoclonal antibody reacts with mouse, rat, and human monocyte chemoattractant protein-1 (MCP-1), also known as CCL2 and MCAF.

          Applications Reported: The 2H5 antibody has been reported for use in ELISA, intracellular staining for flow cytometric analysis, and cytokine neutralization. .

          Applications Tested: The Functional Grade purified 2H5 antibody has been tested by ELISA for verification of reactivity and by LAL assay for verification of low endotoxin. The 2H5 antibody has been published for use in vitro and in vivo for neutralization of MCP-1 activity.

          Mouse MCP-1 ELISA: The biotinylated 2H5 antibody has been tested as the detection antibody in a sandwich ELISA for analysis of mouse MCP-1 in combination with the affinity purified 4E2/MCP (14-7091) antibody for capture and recombinant mouse MCP-1 as the standard. A suitable range of concentrations of this antibody for ELISA detection is 0.5-2.0 µg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 2000 pg/mL - 15 pg/mL should be included in each ELISA plate.

          Human MCP-1 ELISA: The biotinylated 2H5 antibody has been tested as the detection antibody in a sandwich ELISA for analysis of human MCP-1 in combination with the affinity purified 5D3-F7 (14-7099) antibody for capture and recombinant human MCP-1 as the standard. A suitable range of concentrations of this antibody for ELISA detection is 0.5-2.0 µg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 2000 pg/mL - 15 pg/mL should be included in each ELISA plate.

          Storage and handling: Use in a sterile environment.

          Filtration: 0.2 µm post-manufacturing filtered.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Endotoxin Level: Less than 0.001 ng/µg antibody, as determined by LAL assay.

          Aggregation: Less than 10%, as determined by HPLC.

          Target Information

          This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Ccl2; chemokine (C-C motif) ligand 2; chemotactic and activating factor precursor; Genbank Staff Annotation: The small inducible cytokine A2 (SWISS-PROT ACC P13500) is encoded in this mRNA, although it is interrupted and partial, perhaps indicating that this CDS is not the true product of the gamma.1 gene.; H-MCP-1; immediate-early serum-responsive JE protein; immediate-early serum-responsive protein JE; JE; MGC9434; monocyte chemoattractant protein 1; monocyte chemoattractant protein-1; monocyte chemotactic and activating factor; monocyte chemotactic protein; monocyte chemotactic protein 1; monocyte secretory protein JE; Mutations: 171:OK; platelet-derived growth factor-inducible protein JE; RP23-350G1.3; small inducible cytokine A2; small inducible cytokine A2 (monocyte chemotactic protein 1, homologous to mouse Sig-je); small inducible cytokine subfamily A (Cys-Cys), member 2; small inducible gene JE; small-inducible cytokine A2; unnamed protein product

          View more View less

          Gene Aliases: GDCF-2; HC11; HSMCR30; JE; MCAF; MCP-1; MCP1; SCYA2; Sigje; SMC-CF

          View more View less

          UniProt ID: (Human) P13500, (Mouse) P10148, (Rat) P14844

          View more View less

          Entrez Gene ID: (Human) 6347, (Mouse) 20296, (Rat) 24770

          View more View less

          Function(s)
          protein kinase activity receptor binding protein binding chemokine activity CCR2 chemokine receptor binding chemoattractant activity chemokine receptor binding CCR chemokine receptor binding G-protein coupled receptor binding cytokine activity heparin binding
          Process(es)
          angiogenesis response to hypoxia positive regulation of leukocyte mediated cytotoxicity positive regulation of endothelial cell proliferation leukocyte migration involved in inflammatory response chronic inflammatory response monocyte chemotaxis positive regulation of leukocyte migration positive regulation of cellular extravasation cellular calcium ion homeostasis inflammatory response immune response cytoskeleton organization transforming growth factor beta receptor signaling pathway glial cell migration regulation of cell shape response to xenobiotic stimulus response to wounding response to mechanical stimulus response to bacterium response to gamma radiation regulation of vascular endothelial growth factor production positive regulation of gene expression positive regulation of macrophage chemotaxis response to activity negative regulation of angiogenesis cytokine-mediated signaling pathway sensory perception of pain positive regulation of cell-cell adhesion positive regulation of cell migration neutrophil chemotaxis organ regeneration response to lipopolysaccharide response to progesterone positive regulation of tumor necrosis factor production cellular response to insulin stimulus positive regulation of collagen biosynthetic process response to vitamin B3 negative regulation of glial cell apoptotic process response to tumor necrosis factor helper T cell extravasation monocyte homeostasis response to isolation stress cellular response to macrophage colony-stimulating factor stimulus cellular response to platelet-derived growth factor stimulus chemokine (C-C motif) ligand 2 signaling pathway response to amino acid negative regulation of neuron apoptotic process astrocyte cell migration response to ethanol positive regulation of cell adhesion vascular endothelial growth factor receptor signaling pathway eosinophil chemotaxis macrophage chemotaxis lymphocyte chemotaxis positive regulation of synaptic transmission positive regulation of T cell activation positive chemotaxis response to glucocorticoid positive regulation of synaptic transmission, glutamatergic mammary gland involution maternal process involved in female pregnancy maternal process involved in parturition antimicrobial humoral immune response mediated by antimicrobial peptide chemokine-mediated signaling pathway cellular response to lipopolysaccharide cellular response to retinoic acid cellular response to ATP cellular response to glucose stimulus cellular response to interferon-gamma cellular response to interleukin-1 cellular response to interleukin-6 cellular response to tumor necrosis factor cellular response to estradiol stimulus cellular response to fatty acid cellular response to lipoprotein particle stimulus cellular response to high density lipoprotein particle stimulus cellular response to xenobiotic stimulus cellular response to dexamethasone stimulus positive regulation of monocyte chemotaxis positive regulation of immune complex clearance by monocytes and macrophages positive regulation of calcium ion import positive regulation of wound healing positive regulation of protein targeting to membrane energy homeostasis positive regulation of glutamate receptor signaling pathway response to cyclosporin A negative regulation of vascular endothelial cell proliferation negative regulation of G1/S transition of mitotic cell cycle positive regulation of endothelial cell apoptotic process positive regulation of apoptotic cell clearance negative regulation of natural killer cell chemotaxis positive regulation of cytosolic calcium ion concentration osteoclast differentiation chemokinesis cellular response to fibroblast growth factor stimulus protein phosphorylation chemotaxis humoral immune response cell adhesion signal transduction cell surface receptor signaling pathway G-protein coupled receptor signaling pathway G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger JAK-STAT cascade organ morphogenesis viral genome replication cellular homeostasis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-p95z8:80/100.66.74.151:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline