Ácido 3-nitroftálico, 95 %, Thermo Scientific Chemicals
Ácido 3-nitroftálico, 95 %, Thermo Scientific Chemicals
Ácido 3-nitroftálico, 95 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ácido 3-nitroftálico, 95 %, Thermo Scientific Chemicals

CAS: 603-11-2 | C8H5NO6 | 211.13 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
100 g
500 g
2.5 kg
Envase:
Frasco de plástico
Número de catálogo 160161000
Precio (EUR)
51,60
Each
Cantidad:
100 g
Envase:
Frasco de plástico
Pedido a granel o personalizado
Precio (EUR)
51,60
Each
Identificadores químicos
CAS603-11-2
IUPAC Name3-nitrobenzene-1,2-dicarboxylic acid
Molecular FormulaC8H5NO6
InChI KeyKFIRODWJCYBBHY-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(=C1C(O)=O)[N+]([O-])=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Off-white to light yellow
Infrared spectrumConforms
Appearance (Form)Powder
Titration with NaOH>=94 %
HPLC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only