2,4,6-Trimethoxybenzaldehyde, 98%
2,4,6-Trimethoxybenzaldehyde, 98%
2,4,6-Trimethoxybenzaldehyde, 98%
Thermo Scientific Chemicals

2,4,6-Trimethoxybenzaldehyde, 98%

CAS: 830-79-5 | C10H12O4 | 196.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
250 g
Packaging:
Glass bottle
Catalog number 160530250
Price (EUR)
110,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
110,00
Each
Chemical Identifiers
CAS830-79-5
IUPAC Name2,4,6-trimethoxybenzaldehyde
Molecular FormulaC10H12O4
InChI KeyCRBZVDLXAIFERF-UHFFFAOYSA-N
SMILESCOC1=CC(OC)=C(C=O)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Crystalline powder
Melting point118°C to 120°C
Appearance (Color)Beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only