2-Amino-p-cresol, 97%
2-Amino-p-cresol, 97%
2-Amino-p-cresol, 97%
2-Amino-p-cresol, 97%
Thermo Scientific Chemicals

2-Amino-p-cresol, 97%

CAS: 95-84-1 | C7H9NO | 123.16 g/mol
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 163212500
Price (JPY)
-
Request A Quote
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS95-84-1
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Form)Crystalline powder or flakes
Melting point134°C to 139°C
Infrared spectrumConforms
Appearance (Color)Beige to grey-brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only