3-Methoxy-2(1H)-pyridone, 97%
3-Methoxy-2(1H)-pyridone, 97%
3-Methoxy-2(1H)-pyridone, 97%
Thermo Scientific Chemicals

3-Methoxy-2(1H)-pyridone, 97%

CAS: 20928-63-6 | C6H7NO2 | 125.13 g/mol
Quantity:
1 g
5 g
Packaging:
View all
Glass bottle
Catalog number 163540010
Price (JPY)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS20928-63-6
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Powder
Appearance (Color)White to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only