1,3-Dicyanobenzene, 98%
1,3-Dicyanobenzene, 98%
1,3-Dicyanobenzene, 98%
Thermo Scientific Chemicals

1,3-Dicyanobenzene, 98%

CAS: 626-17-5 | C8H4N2 | 128.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
Packaging:
View all
Plastic bottle
Catalog number 163662500
Price (USD)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS626-17-5
IUPAC Namebenzene-1,3-dicarbonitrile
Molecular FormulaC8H4N2
InChI KeyLAQPNDIUHRHNCV-UHFFFAOYSA-N
SMILESN#CC1=CC(=CC=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to tan
Melting point159°C to 163°C
Appearance (Form)Flakes
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only