Potassium tert-pentoxide, 0.9M (14 wt.%) solution in cyclohexane, AcroSeal™
Potassium tert-pentoxide, 0.9M (14 wt.%) solution in cyclohexane, AcroSeal™
Potassium tert-pentoxide, 0.9M (14 wt.%) solution in cyclohexane, AcroSeal™
Potassium tert-pentoxide, 0.9M (14 wt.%) solution in cyclohexane, AcroSeal™
Thermo Scientific Chemicals

Potassium tert-pentoxide, 0.9M (14 wt.%) solution in cyclohexane, AcroSeal™

CAS: 41233-93-6 | C5H11KO | 126.24 g/mol
数量:
100 mL
800 mL
パッケージ:
すべて表示
AcroSeal™ Glass bottle
製品番号(カタログ番号) 164721000
価格(JPY)
-
数量:
100 mL
パッケージ:
AcroSeal™ Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS41233-93-6
IUPAC Namepotassium 2-methylbutan-2-olate
Molecular FormulaC5H11KO
InChI KeyZRLVQFQTCMUIRM-UHFFFAOYSA-N
SMILES[K+].CCC(C)(C)[O-]
さらに表示
Appearance (Color)Colorless to yellow
Titration with HClO40.85 to 1.10 M
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only