2-Methylcyclohexanol, 99%, mixture of cis and trans
2-Methylcyclohexanol, 99%, mixture of cis and trans
2-Methylcyclohexanol, 99%, mixture of cis and trans
Thermo Scientific Chemicals

2-Methylcyclohexanol, 99%, mixture of cis and trans

CAS: 583-59-5 | C7H14O | 114.19 g/mol
数量:
250 g
1 kg
2.5 kg
製品番号(カタログ番号) 166890010
価格(JPY)
-
数量:
1 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS583-59-5
IUPAC Name2-methylcyclohexan-1-ol
Molecular FormulaC7H14O
InChI KeyNDVWOBYBJYUSMF-UHFFFAOYNA-N
SMILESCC1CCCCC1O
さらに表示
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4610 to 1.4650 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only