6-Chloronicotinic acid, 99%
6-Chloronicotinic acid, 99%
6-Chloronicotinic acid, 99%
Thermo Scientific Chemicals

6-Chloronicotinic acid, 99%

CAS: 5326-23-8 | C6H4ClNO2 | 157.56 g/mol
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 168691000
Price (JPY)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5326-23-8
IUPAC Name6-chloropyridine-3-carboxylic acid
Molecular FormulaC6H4ClNO2
InChI KeyUAWMVMPAYRWUFX-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(Cl)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point186°C to 190°C
HPLC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only