1,3-Diacetylbenzene, 99%, Thermo Scientific Chemicals
1,3-Diacetylbenzene, 99%, Thermo Scientific Chemicals
1,3-Diacetylbenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Diacetylbenzene, 99%, Thermo Scientific Chemicals

CAS: 6781-42-6 | Molecular Formula: C10H10O2 | Molecular Weight: 162.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1700900505 gGlass bottle
Catalog number 170090050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6781-42-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
Melting point28°C to 33°C
GC>=98.5 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only