Dimethyl-d6 sulfate, for NMR, 99+ atom % D
Dimethyl-d6 sulfate, for NMR, 99+ atom % D
Dimethyl-d6 sulfate, for NMR, 99+ atom % D
Thermo Scientific Chemicals

Dimethyl-d6 sulfate, for NMR, 99+ atom % D

CAS: 15199-43-6 | C2H6O4S | 132.163 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1729100101 gGlass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 172910010
Price (EUR)
-
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS15199-43-6
IUPAC Namedi(²H₃)methyl sulfate
Molecular FormulaC2H6O4S
InChI KeyVAYGXNSJCAHWJZ-WFGJKAKNSA-N
SMILES[2H]C([2H])([2H])OS(=O)(=O)OC([2H])([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Water=<0.05 %
Appearance (Color)Clear colorless
D-Enrichment>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only