1,3-acetonedicarboxylic acid, 96%
1,3-acetonedicarboxylic acid, 96%
1,3-acetonedicarboxylic acid, 96%
Thermo Scientific Chemicals

1,3-acetonedicarboxylic acid, 96%

CAS: 542-05-2 | C5H6O5 | 146.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Catalog number 173151000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS542-05-2
IUPAC Name3-oxopentanedioate
Molecular FormulaC5H4O5
InChI KeyOXTNCQMOKLOUAM-UHFFFAOYSA-L
SMILES[O-]C(=O)CC(=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Powder or crystals or crystalline powder
Infrared spectrumConforms
Titration with NaOH95 to 105 %
Sulfuric acid=<2 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only